Skip to Content
Merck
All Photos(1)

Key Documents

733113

Sigma-Aldrich

N-Methyldiethanol-1,1,1′,1′-d4 amine

98 atom % D, 97% (CP)

Synonym(s):

2,2′-Methyliminodiethanol-1,1,1′,1′-d4, MDEA-1,1,1′,1′-d4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3N(CH2CD2OH)2
Molecular Weight:
123.19
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

97% (CP)

form

liquid

bp

246-248 °C (lit.)

density

1.072 g/mL at 25 °C

mass shift

M+4

SMILES string

[2H]C([2H])(O)CN(C)CC([2H])([2H])O

InChI

1S/C5H13NO2/c1-6(2-4-7)3-5-8/h7-8H,2-5H2,1H3/i4D2,5D2

InChI key

CRVGTESFCCXCTH-CQOLUAMGSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service