Skip to Content
Merck
All Photos(1)

Key Documents

714216

Sigma-Aldrich

2-Deoxystreptamine dihydrobromide

≥97% (TLC)

Synonym(s):

2-DOS · 2HBr, 4t,6t-Diamino-cyclohexan-1r,2t,3c-triol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O3 · 2HBr
CAS Number:
Molecular Weight:
324.01
Beilstein:
3692825
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97% (TLC)

form

solid

storage temp.

2-8°C

SMILES string

Br.Br.N[C@H]1C[C@@H](N)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H14N2O3.2BrH/c7-2-1-3(8)5(10)6(11)4(2)9;;/h2-6,9-11H,1,7-8H2;2*1H/t2-,3+,4+,5-,6-;;

InChI key

FQPOWLZRZNUTOR-SKIYRPIFSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sicong Li et al.
ACS chemical biology, 13(8), 2203-2210 (2018-06-08)
Hygromycin B is an aminoglycoside antibiotic with a structurally distinctive orthoester linkage. Despite its long history of use in industry and in the laboratory, its biosynthesis remains poorly understood. We show here, by in-frame gene deletion in vivo and detailed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service