Skip to Content
Merck
All Photos(1)

Key Documents

711713

Sigma-Aldrich

1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide

≥98%

Synonym(s):

BMIIm, BMIM TFSI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15F6N3O4S2
CAS Number:
Molecular Weight:
419.36
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

liquid

impurities

≤0.5% water

functional group

fluoro

SMILES string

CCCCn1cc[n+](C)c1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C8H15N2.C2F6NO4S2/c1-3-4-5-10-7-6-9(2)8-10;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h6-8H,3-5H2,1-2H3;/q+1;-1

InChI key

INDFXCHYORWHLQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sales restrictions may apply

Application

1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide is a hydrophobic ionic liquid that can enhance the activity of N,N,N′,N′-tetra(n-octyl)diglycolamide (TODGA) extractant in the extraction of uranium(VI) from an aqueous nitric acid media.

Caution

not available in the US

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synergistic extraction of uranium (VI) with TODGA and hydrophobic ionic liquid mixtures into molecular diluent.
Turanov AN, et al.
Separation and Purification Technology, 164, 97-106 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service