Skip to Content
Merck
All Photos(2)

Key Documents

680400

Sigma-Aldrich

4,4′-Diformyltriphenylamine

95%

Synonym(s):

4,4′-(Phenylimino)bisbenzaldehyde, Bis(4-formylphenyl)aniline, Bis(4-formylphenyl)phenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H15NO2
CAS Number:
Molecular Weight:
301.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

142-146 °C

functional group

aldehyde

SMILES string

[H]C(=O)c1ccc(cc1)N(c2ccccc2)c3ccc(cc3)C([H])=O

InChI

1S/C20H15NO2/c22-14-16-6-10-19(11-7-16)21(18-4-2-1-3-5-18)20-12-8-17(15-23)9-13-20/h1-15H

InChI key

DOUAFMIJGIUWJX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Viprabha Kakekochi et al.
ChemPlusChem, 85(8), 1762-1777 (2020-08-15)
A set of four symmetric, butterfly-shaped 4-(4-(decyloxy)phenyl)-2,6-di(thiophen-2-yl)pyridine (TPY) derivatives 2TPA-TPY (TPY center and triphenylamine end groups), 2CBZ-TPY (TPY center and N-ethyl carbazole end groups), 2TPY-TPA (triphenylamine center and TPY at the periphery) and 2TPY-CBZ (N-ethyl carbazole center and TPY at
Patricio A Sobarzo et al.
Polymers, 11(2) (2019-04-10)
Three new triphenylamine-based oligomeric Schiff bases (polySB1, polySB2 and polySB3) containing tetraphenylsilane core (TPS-core) in the main chain were obtained from TPS-core-based diamines and bis(4-formylphenyl)phenylamine by a high-temperature polycondensation reaction. These new oligomers were structurally characterized by FT-IR, NMR and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service