676144
Sodium D-lactate-3-13C solution
45-55% in H2O, 99 atom % 13C
Synonym(s):
D-Lactic -3-13C acid sodium salt solution
About This Item
Recommended Products
description
Packaged as gram of solution, 45-55%(w/w)
isotopic purity
99 atom % 13C
concentration
45-55% in H2O
technique(s)
mass spectrometry (MS): suitable
mass shift
M+1
SMILES string
[Na+].[13CH3][C@@H](O)C([O-])=O
InChI
1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m1./s1/i1+1;
InChI key
NGSFWBMYFKHRBD-MDBPNCOTSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Packaging
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service