660264
2-Butene-1,1,1-d3, mixture of cis and trans
99 atom % D, 98% (CP)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH=CHCD3
Molecular Weight:
59.12
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
Recommended Products
isotopic purity
99 atom % D
Assay
98% (CP)
form
gas
bp
1 °C
mass shift
M+3
SMILES string
[2H]C([2H])([2H])\C=C\C
InChI
1S/C4H8/c1-3-4-2/h3-4H,1-2H3/b4-3+/i1D3
InChI key
IAQRGUVFOMOMEM-VGIDZVCYSA-N
Related Categories
Packaging
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Gas 1 - Press. Gas Liquefied gas
Storage Class Code
2A - Gases
WGK
nwg
Flash Point(F)
-4.0 °F - closed cup
Flash Point(C)
-20 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service