Skip to Content
Merck
All Photos(2)

Key Documents

608629

Sigma-Aldrich

Glyphosate-2-13C,15N

98 atom % 15N, 99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C13CH215NHCH2PO3H2
CAS Number:
Molecular Weight:
171.06
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C
98 atom % 15N

Quality Level

mp

230 °C (dec.) (lit.)

application(s)

agriculture
environmental

mass shift

M+2

SMILES string

OC(=O)[13CH2][15NH]CP(O)(O)=O

InChI

1S/C3H8NO5P/c5-3(6)1-4-2-10(7,8)9/h4H,1-2H2,(H,5,6)(H2,7,8,9)/i1+1,4+1

InChI key

XDDAORKBJWWYJS-VFZPYAPFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glyphosate-2-13C,15N is enriched with 13C and 15N isotopes.

Application

Glyphosate-2-13C,15N has been used widely in the field of agriculture. It is also used as an internal standard for water quality determination.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lene Nørby Nielsen et al.
Environmental pollution (Barking, Essex : 1987), 233, 364-376 (2017-11-03)
Recently, concerns have been raised that residues of glyphosate-based herbicides may interfere with the homeostasis of the intestinal bacterial community and thereby affect the health of humans or animals. The biochemical pathway for aromatic amino acid synthesis (Shikimate pathway), which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service