Skip to Content
Merck
All Photos(1)

Key Documents

606219

Sigma-Aldrich

Erythromycin-(N,N-dimethyl-13C2)

99 atom % 13C, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C2C35H67NO13
CAS Number:
Molecular Weight:
735.91
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

95% (CP)

form

solid

mass shift

M+2

SMILES string

CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N([13CH3])[13CH3])[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O

InChI

1S/C37H67NO13/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3/t18-,19-,20+,21+,22-,23+,24+,25-,26+,28-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1/i11+1,12+1

InChI key

ULGZDMOVFRHVEP-AXHSKANISA-N

Related Categories

General description

Erythromycin- (N, N-dimethyl-13C2) is an isotopic analogue of Erythromycin-A

Application

Erythromycin-(N, N-dimethyl-13C2) can be used as an internal standard for the quantitative analysis of erythromycin residues. It is also used to study the uptake and depletion time of erythromycin in natural proportion after oral administration of the drug.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dario Lucchetti et al.
Journal of agricultural and food chemistry, 53(25), 9689-9694 (2005-12-08)
In recent years, erythromycin has received considerable attention for its therapeutic efficacy against some bacterial kidney diseases in aquaculture and, therefore, suitable and sensitive analytical methods to monitor erythromycin residues in fish are required. A fast sample treatment followed by
Annarita Esposito et al.
Antimicrobial agents and chemotherapy, 51(3), 1043-1047 (2006-12-30)
Aquaculture production has notably increased in the last decades, mainly thanks to intensive farming. Together with market globalization, this gives rise to the spreading of several fish diseases, thus increasing the demand for veterinary drugs for aquatic species. Nonetheless, very

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service