Skip to Content
Merck
All Photos(1)

Key Documents

488879

Sigma-Aldrich

Dimethyl ether-d6

99 atom % D, 99% (CP)

Synonym(s):

Methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CD3)2O
CAS Number:
Molecular Weight:
52.11
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

vapor density

1.62 (vs air)

Quality Level

vapor pressure

7760 mmHg ( 25 °C)

isotopic purity

99 atom % D

Assay

99% (CP)

form

gas

autoignition temp.

662 °F

bp

−24.8 °C (lit.)

mp

−141 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])OC([2H])([2H])[2H]

InChI

1S/C2H6O/c1-3-2/h1-2H3/i1D3,2D3

InChI key

LCGLNKUTAGEVQW-WFGJKAKNSA-N

Related Categories

Packaging

5 g quantitiy is packaged in a 25 mL Sure/Pac cylinder with brass 1/4" male NPT valve. Nominal gas pressure at 21ºC is 60 psig for 5 g. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Legal Information

Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service