Skip to Content
Merck

195359

Sigma-Aldrich

4-Fluorobenzotrifluoride

98%

Synonym(s):

α,α,α,4-Tetrafluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CF3
CAS Number:
Molecular Weight:
164.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.401 (lit.)

bp

102-105 °C (lit.)

mp

−42-−41.7 °C (lit.)

density

1.293 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Fc1ccc(cc1)C(F)(F)F

InChI

1S/C7H4F4/c8-6-3-1-5(2-4-6)7(9,10)11/h1-4H

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB4301192HPA008310HPA068103
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

conjugate

unconjugated

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

-

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

Application

4-Fluorobenzotrifluoride was used in the synthesis of:
  • 2,2′-bis(trifluoromethylphenoxy)biphenyl via nucleophilic aromatic substitution reaction with 2,2′-biphenol[1]
  • (S)-fluoxetine, potent inhibitor of neuronal serotonin-uptake[2]
  • 1-aryloxy-2-substituted aminomethyltetrahydronaphthalene derivatives[3]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kalpana Bhandari et al.
Bioorganic & medicinal chemistry, 14(8), 2535-2544 (2005-12-13)
Several 1-aryloxy-2-substituted aminomethyltetrahydronaphthalenes (7-21) as conformationally rigid analogues of fluoxetine were synthesized and evaluated for their anorexigenic and antidepressant activities. For SAR studies the related acyclic analogues (22-27) were also prepared. Out of the 21 synthesized compounds, 10 compounds (9
Akiko Okamoto et al.
Journal of oleo science, 56(9), 479-491 (2007-09-28)
Novel bifunctional acyl-acceptant biphenyls bearing trifluoromethylated aroyloxy groups were successfully synthesized in high yields via TfOH-mediated electrophilic aromatic aroylation of fluorobenzene with CF(3)-bearing aroyl chlorides followed by nucleophilic aromatic substitution with 2,2'-biphenol. Similarly, 2,2'-bis(trifluoromethylphenoxy)biphenyl was synthesized via nucleophilic aromatic substitution
D W Robertson et al.
Journal of medicinal chemistry, 31(7), 1412-1417 (1988-07-01)
Fluoxetine is a potent and selective inhibitor of the neuronal serotonin-uptake carrier and is a clinically effective antidepressant. Although fluoxetine is used therapeutically as the racemate, there appears to be a small but demonstrable stereospecificity associated with its interactions with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service