Merck
  • Somatic cancer mutations in the DNMT2 tRNA methyltransferase alter its catalytic properties.
Somatic cancer mutations in the DNMT2 tRNA methyltransferase alter its catalytic properties.
Biochimie (2015-03-10)
Winfried Elhardt, Raghuvaran Shanmugam, Tomasz P Jurkowski, Albert Jeltsch
ABSTRACT
Methylation of tRNA is an important post-transcriptional modification and aberrations in tRNA modification has been implicated in cancer. The DNMT2 protein methylates C38 of tRNA-Asp and it has a role in cellular physiology and stress response and its expression levels are altered in cancer tissues. Here we studied whether DNMT2 somatic mutations found in cancer tissues affect the activity of the enzyme. We have generated 13 DNMT2 variants and purified the corresponding proteins. All proteins were properly folded as determined by circular dichroism spectroscopy. We tested their RNA methylation activity using in vitro generated tRNA-Asp. One of the mutations (E63K) caused a twofold increase in activity, while two of them led to a strong (over fourfold) decrease in activity (G155S and L257V). Two additional mutant proteins were almost inactive (R371H and G155V). The strong effect of some of the somatic cancer mutations on DNMT2 activity suggests that these mutations have a functional role in tumorigenesis.
Sigma-Aldrich
DNMT2 Active human, recombinant, expressed in baculovirus infected insect cells, ≥80% (SDS-PAGE)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Pricing and availability is not currently available.
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Pricing and availability is not currently available.