Merck
  • Synthesis and antibacterial activity of some novel 4-oxopyrido[2,3-a]phenothiazines.
Synthesis and antibacterial activity of some novel 4-oxopyrido[2,3-a]phenothiazines.
Archiv der Pharmazie (2014-09-16)
Hadeel T Al-Sinjilawi, Mustafa M El-Abadelah, Mohammad S Mubarak, Amal Al-Aboudi, Mohammed M Abadleh, Adel M Mahasneh, Asaad K M A Ahmad
ABSTRACT
A series of substituted 4-oxopyrido[2,3-a]phenothiazine-3-carboxylic acids (6a-d) were prepared via cyclization of the corresponding ethyl 7-(arylthioxy)-8-nitro(or azido)-4-oxoquinoline-3-carboxylates (3a-d/4a-d), followed by hydrolysis of the resultant esters (5a-d). Among these tetracyclics, compound 6a with unsubstituted terminal benzo-ring D was the most active against representative Gram-positive and Gram-negative bacterial strains. These compounds were also active against methicillin-resistant Staphylococcus aureus (MRSA), with very low toxicity to normal cells. Virtual screening using ligand-protein docking modeling predicted that the compounds 6a-d are potential inhibitors of the topoisomerase IV enzyme and that hydrophobic interactions and hydrogen bonds are the major molecular interactions between these compounds and the residues of the active site of topoisomerase IV.
Sigma-Aldrich
Thiophenol, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Cyclopropylamine, 98%
Pricing and availability is not currently available.
Supelco
Thiophenol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydride, dry, 90%
Pricing and availability is not currently available.
Sigma-Aldrich
Thiophenol, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Chlorothiophenol, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
4-Fluorothiophenol, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Methylbenzenethiol, 98%
Pricing and availability is not currently available.