Skip to Content
Merck
Micro particles based on melamine resin, FITC-marked size: 6 μm

Micro particles based on melamine resin, FITC-marked

Micro particles based on melamine resin, FITC-marked size: 6 μm
Synonym(s):
Melamine-formaldehyde resin

Products

Product Category
Brand
Physical Form
Format
Quality Segment
Application
Particle Size (µm)
Reference Material Type
Special Grade