Skip to Content
Merck
All Photos(1)

Key Documents

V900414

Sigma-Aldrich

Sulfamethazine

Vetec, reagent grade, ≥99%

Synonym(s):

4,6-Dimethylsulfadiazine, 4-Amino-N-(4,6-dimethyl-2-pyrimidinyl)benzenesulfonamide, Sulfadimethyldiazine, Sulfadimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N4O2S
CAS Number:
Molecular Weight:
278.33
Beilstein:
261304
EC Number:
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥99%

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

storage temp.

2-8°C

SMILES string

Cc1cc(C)nc(NS(=O)(=O)c2ccc(N)cc2)n1

InChI

1S/C12H14N4O2S/c1-8-7-9(2)15-12(14-8)16-19(17,18)11-5-3-10(13)4-6-11/h3-7H,13H2,1-2H3,(H,14,15,16)

InChI key

ASWVTGNCAZCNNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

An antimicrobial sulfur drug. Induces CYP3A4 expression and acetylated by N-acetyltransferase. Exhibits sex dependent pharmacokinetics, metabolized by the male specific isoform CYP2C11.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ma Jesús García-Galán et al.
The Science of the total environment, 409(24), 5505-5512 (2011-09-29)
Degradation of the sulfonamide sulfamethazine (SMZ) by the white-rot fungus Trametes versicolor was assessed. Elimination was achieved to nearly undetectable levels after 20 h in liquid medium when SMZ was added at 9 mg L(-1). Experiments with purified laccase and
M Rérat et al.
Preventive veterinary medicine, 103(4), 265-273 (2011-09-29)
The present study was conducted to evaluate the efficacy of two prophylactic antibiotic treatments against bovine respiratory disease (BRD) in veal calves. In addition, the antibiotic susceptibilities of isolated Pasteurellaceae were tested. The calves were treated either on the day
Vincent Curtin et al.
Molecular pharmaceutics, 10(1), 386-396 (2012-11-29)
The coprocessing of active pharmaceutical ingredient (API) with an excipient which has a high glass transition temperature (T(g)) is a recognized strategy to stabilize the amorphous form of a drug. This work investigates whether coprocessing a model API, sulfadimidine (SDM)
Marc Teixidó et al.
Environmental science & technology, 45(23), 10020-10027 (2011-10-27)
Adsorption of ionizable compounds by black carbon is poorly characterized. Adsorption of the veterinary antibiotic sulfamethazine (SMT; a.k.a., sulfadimidine; pK(a1) = 2.28, pK(a2) = 7.42) on a charcoal was determined as a function of concentration, pH, inorganic ions, and organic
Marina Islas-Espinoza et al.
Microbial ecology, 64(1), 140-151 (2012-01-31)
Persistence or degradation of synthetic antibiotics in soil is crucial in assessing their environmental risks. Microbial catabolic activity in a sandy loamy soil with pig manure using 12C- and 14C-labelled sulfamethazine (SMZ) respirometry showed that SMZ was not readily degradable.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service