About This Item
Recommended Products
grade
AR
product line
Vetec™
Assay
99-100.5%
form
crystalline powder
pH
5-8 (5% in water)
cation traces
Fe: ≤0.0002%
heavy metals (as Pb): ≤0.0005%
SMILES string
O.O.Cl[Ba]Cl
InChI
1S/Ba.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2
InChI key
PWHCIQQGOQTFAE-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service