Skip to Content
Merck
All Photos(1)

Key Documents

1A02750

USP

3-(1-Cyanoethyl)benzoic acid

Pharmaceutical Analytical Impurity (PAI)

Synonym(s):

2-(3-Carboxyphenyl)Propionitrile

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€839.00

€839.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€839.00

About This Item

Linear Formula:
CH3CH(CN)C6H4CO2H
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.24

€839.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical analytical impurity (PAI)

Agency

USP

manufacturer/tradename

USP

mp

145-148 °C (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

CC(C#N)c1cccc(c1)C(O)=O

InChI

1S/C10H9NO2/c1-7(6-11)8-3-2-4-9(5-8)10(12)13/h2-5,7H,1H3,(H,12,13)

InChI key

IRYIYPWRXROPSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(1-Cyanoethyl)benzoic acid is a USP Pharmaceutical Analytical Impurity (PAI).

USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.

Associated Drug Substance: Ketoprofen.

For more information about this PAI, visit here.

Application

3-(1-Cyanoethyl)benzoic acid (USP PAI) is intended for use in analytical testing to detect, identify, and measure pharmaceutical impurities.

Features and Benefits

USP PAI advance your early analytical R&D and process development. PAI can be used in the following applications:
1. Conduct analytical tests during early formulation feasibility studies.
2. Determine degradation impurities produced during stress studies.
3. Develop, validate, and transfer analytical methods.
4. Perform spiking studies during process R&D to demonstrate depletion upon recrystallization.
5. Record retention times and/or spectra
6. Determine relative response factors.
7. Identify unknown impurities that formed during ICH stability conditions.
8. Identify impurities that are present in the Reference Listed Drug
9. Test for and profile impurities not listed in drug substance and drug product monographs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service