Skip to Content
Merck
All Photos(1)

Documents

G006166

LC/MS Analysis of 2C Amines Drugs of Abuse on Ascentis® Express HILIC

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
mobile phase: 5 mM ammonium formate in acetonitrile:water (90:10, v/v)
flow rate: 0.4 mL/min
column temp.: 35 °C
detector: TOF/MS
detector: 2.0 μL
sample: 300 ng/mL in acetonitrile

suitability

application for HPLC

application(s)

clinical
forensics and toxicology

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    (R)-(-)-Bromo-DragonFLY hydrochloride solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    (±)-2,5-Dimethoxy-4-Iodoamphetamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    (±)-2,5-Dimethoxy-4-bromoamphetamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    2C-B-FLY hydrochloride solution, 1 mg/mL in acetonitrile: water (9:1), ampule of 1 mL, certified reference material, Cerilliant®

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service