Skip to Content
Merck
All Photos(1)

Key Documents

G005482

LC/MS Analysis of Lactic Acid Enantiomers in Urine on Astec® CHIROBIOTIC® R

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

technique(s)

HPLC: suitable

test parameters

column: Astec CHIROBIOTIC R, 15 cm x 2.1 mm I.D., 5 μm particles (13019AST)
mobile phase: [A] 33.3 mM ammonium acetate; [B] acetonitrile; (15:85, A:B)
flow rate: 0.7 mL/min
column temp.: 4 °C
detector: (-)ESI
injection: 20 μL
sample: Urine extracts in 5% (v/v) acetic acid, 10% (v/v) H2O and 85% (v/v) acetonitrile

suitability

application for HPLC

application(s)

clinical

Analysis Note

Chromatogram supplied courtesy of H. Henry and associates at the Clinical Chemistry Laboratory, Centre Hospitalier Universitaire Vaudois, University of Lausanne, CH-1011, Switzerland. View the related full-length contributed article in Reporter 30.1.

Other Notes

Reference: Biomed. Chromatogr. 2011 "Sensitive determination of D-lactic acid and L-lactic acid in urine by high-performance liquid chromatography–tandem mass spectrometry" H. Henry, N. Marmy Conus, P. Steenhout, A. Béguin and O. Boulat, Published online in Wiley Online Library, wileyonlinelibrary.com/journal/bmc.

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    L-(+)-Lactic acid, analytical standard
    D-(−)-Lactic acid, ≥90%

mobile phase component

Product No.
Description
Pricing

standard

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service