Skip to Content
Merck
All Photos(1)

Documents

G005173

SFC Analysis of Flavanone Enantiomers on Astec® Cellulose DMP, Methanol Additive

application for SFC

Sign Into View Organizational & Contract Pricing


About This Item

test parameters

column: Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase: [A] CO2; [B] isopropanol; (80:20, A:B)
flow rate: 3 mL/min
pressure: 1450 psi (100 bar)
column temp.: 35 °C
detector: UV, 220 nm
injection: 5 μL
sample: flavanone, 2 mg/mL

suitability

application for SFC

application(s)

vitamins, nutraceuticals, and natural products

Analysis Note

SFC data provided by Dr. Christina Kraml, Lotus Separations, LLC, Princeton, NJ.
Instrument: Berger Analytical SFC system equipped with a dual pump (FCM-1200), autosampler (ALS 3100), column oven (TCM-200) and diode-array detector (DAD-4100); from Mettler Toledo.

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Flavanone, 98%

standard

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service