Skip to Content
Merck
All Photos(1)

Key Documents

816095

TSKgel® Phenyl-5PW (20 µm) HPLC Columns

L x I.D. 3.5 cm × 10 mm, phenyl phase

Synonym(s):

TSKgel® Phenyl-5PW HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€1,390.00

€1,390.00


Please contact Customer Service for Availability


Select a Size

Change View
1 KIT
€1,390.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

€1,390.00


Please contact Customer Service for Availability

Product Name

TSKgel® Phenyl-5PW Guardgel Kit, phase phenyl, L × I.D. 3.5 cm × 10 mm, 20 μm particle size

material

stainless steel

product line

TSKgel®

manufacturer/tradename

Tosoh Phenyl-5PW, 16095

parameter

≤4 M salt concentration
≤50% organic solvent
10-50 °C temperature

technique(s)

HPLC: suitable

L × I.D.

3.5 cm × 10 mm

matrix

polymer particle platform

matrix active group

phenyl phase

particle size

20 μm

pore size

1000 Å

operating pH

2-12

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

Guardgel Kit for 21.5mmID TSKgel Phenyl-5PW columns

Components

TSKgel Guardgel kits for 21.5 mm ID preparative columns contain:
  • Column Holder, consisting of external reducing union, cap, frit, 2 ferrules
  • Column, 35mm x 12.75mm OD x 10mm ID, 10μm frit at 20mm depth, 1/16"
  • 2 Replacement frits
  • Capillary Tubing, 5cm x 1/16" OD x 0.02" ID
  • Nuts and Ferrules, 10 - 32 threads for 1/16" tubing, 2 each
  • 2 x 5mL guardgel

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service