Skip to Content
Merck
All Photos(1)

Key Documents

73317

Supelco

1H,1H,2H,2H-Perfluorohexan-1-ol

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol

Sign Into View Organizational & Contract Pricing

Select a Size

73317-25MG
€134.00

€134.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
73317-25MG
€134.00

About This Item

Linear Formula:
CF3(CF2)3(CH2)2OH
CAS Number:
Molecular Weight:
264.09
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€134.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

LC/MS: suitable

refractive index

n20/D 1.314 (lit.)

bp

140-143 °C (lit.)

density

1.59 g/mL at 25 °C (lit.)

application(s)

clinical testing
coatings
environmental
food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C6H5F9O/c7-3(8,1-2-16)4(9,10)5(11,12)6(13,14)15/h16H,1-2H2

InChI key

JCMNMOBHVPONLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.9 °F - closed cup

Flash Point(C)

73.3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service