Skip to Content
Merck
All Photos(1)

Key Documents

72799

Supelco

Bis(2,2,6,6-tetramethyl-4-piperidyl) sebacate

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€64.40

€64.40


Please contact Customer Service for Availability


Select a Size

Change View
100 MG
€64.40

About This Item

Empirical Formula (Hill Notation):
C28H52N2O4
CAS Number:
Molecular Weight:
480.72
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.24

€64.40


Please contact Customer Service for Availability

grade

analytical standard

Quality Level

Assay

≥97.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

82-85 °C (lit.)

transition temp

Tg >350 °C (dec)

format

neat

storage temp.

2-30°C

SMILES string

CC1(C)CC(CC(C)(C)N1)OC(=O)CCCCCCCCC(=O)OC2CC(C)(C)NC(C)(C)C2

InChI

1S/C28H52N2O4/c1-25(2)17-21(18-26(3,4)29-25)33-23(31)15-13-11-9-10-12-14-16-24(32)34-22-19-27(5,6)30-28(7,8)20-22/h21-22,29-30H,9-20H2,1-8H3

InChI key

XITRBUPOXXBIJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>302.0 °F

Flash Point(C)

> 150 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service