52171-U
SPE Tube Cap (encloses top of SPE tubes)
for use with 1mL SPE tube, pk of 108
Synonym(s):
PK108 CAPS FOR 1ML SPE TUBES
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.21
Recommended Products
material
yellow polypropylene
description
Plugs top of SPE tubes
packaging
pk of 108
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
compatibility
for use with 1mL SPE tube
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service