Key Documents
502472
MicroTee
for use with MicroTight Sleeves, pkg of 1 ea
About This Item
UNSPSC Code:
41121815
description
Connects 3 pieces of 0.025″ O.D. capillary tubing
includes 3, microferrules for 0.025″ O.D. tubing and 3, female nuts (5/16"-24 internal threads)
packaging
pkg of 1 ea
compatibility
for use with 0.025″ O.D. tubing
for use with MicroTight Sleeves
General description
Connects 3 capillary tubes, using MicroFingertight II fittings. Includes 3 MicroFerrules for 0.025 in. O.D. tubing, 3 female nuts.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)