Skip to Content
Merck
All Photos(1)

Key Documents

48487-U

Supelco

Aflatoxin B1, B2, G1, G2 Mix

Certified Reference Material, Varied Concentration

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available. Contact Technical Service

grade

certified reference material

CofA

current certificate can be downloaded

packaging

ampule of 3 mL

storage temp.

2-8°C

Analyte

Description

    Aflatoxin G2
    Aflatoxin G1
    Aflatoxin B2
    Aflatoxin B1

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Flam. Liq. 3 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service