Skip to Content
Merck
All Photos(1)

Key Documents

47182

Supelco

o-Tolualdehyde-2,4-DNPH solution

100 μg/mL in acetonitrile (as aldehyde), analytical standard

Synonym(s):

o-Tolualdehyde-2,4-dinitrophenylhydrazone solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12000000

grade

analytical standard

CofA

certificate of analysis is enclosed in each package.

feature

standard type calibration

packaging

ampule of 1 mL

concentration

100 μg/mL in acetonitrile (as aldehyde)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C14H12N4O4/c1-10-4-2-3-5-11(10)9-15-16-13-7-6-12(17(19)20)8-14(13)18(21)22/h2-9,16H,1H3/b15-9+

InChI key

QUIFNJPMGQCNMH-OQLLNIDSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This product is now available as a certified reference material. Please reference CRM4M7182.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service