Skip to Content
Merck
All Photos(2)

Key Documents

44894-U

Supelco

ASTM® D6584 1,3-Diolein Solution

5000 μg/mL in pyridine, analytical standard

Synonym(s):

Biodiesel impurity

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Agency

ASTM® D6584

form

liquid

CofA

current certificate can be downloaded

packaging

pkg of 2 mL

concentration

5000 μg/mL in pyridine

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

single component solution

functional group

ester

storage temp.

2-8°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)COC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C39H72O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-35-37(40)36-44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,37,40H,3-16,21-36H2,1-2H3/b19-17-,20-18-

InChI key

DRAWQKGUORNASA-CLFAGFIQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhang-Qun Duan et al.
Journal of biotechnology, 159(1-2), 44-49 (2012-03-01)
t-Butanol was an excellent reaction medium for enzyme-mediated esterification of oleic acid with glycerol for 1,3-diolein preparation which has been proved by our group. Herein, to achieve the rational synthesis of 1,3-diolien, response surface methodology was applied to examine the
Zhang-Qun Duan et al.
Bioresource technology, 102(23), 11048-11050 (2011-10-08)
We investigated the influence of solvent on the positional selectivity of Novozym 435 which was the immobilized Candida antarctica lipase B (CALB) during the esterification of oleic acid with glycerol for 1,3-diolein preparation previously. Herein, molecular modeling was used to
G Glowacz et al.
Chemistry and physics of lipids, 79(2), 101-106 (1996-03-29)
The stereoselectivity of porcine pancreas lipase (PPL) was investigated during the enzymatic hydrolysis of triolein and its partial glycerides in the presence of supercritical carbon dioxide (SCCO2) as reaction medium. The water content of the immobilized lipases was varied. The
Zhang-Qun Duan et al.
Bioresource technology, 101(7), 2568-2571 (2009-12-22)
The influence of solvents with a wide range of log P (from -0.23 to 4.5) on the positional selectivity of the immobilized lipase Novozym 435 during the esterification of oleic acid with glycerol for 1,3-diolein preparation was investigated. Analysis was
B Bastide et al.
Experimental cell research, 214(2), 519-527 (1994-10-01)
Possible regulatory effects on cell-to-cell communication of a synthetic diacylglycerol, an activator of protein kinase C (PKC), were examined in pairs of synchronously beating ventricular myocytes of neonatal rats in primary culture. Junctional communication was estimated by measuring either the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service