Skip to Content
Merck
All Photos(1)

Key Documents

442272

Supelco

1-Octadecene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15CH=CH2
CAS Number:
Molecular Weight:
252.48
Beilstein:
1761067
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

0.71 (vs air)

autoignition temp.

482 °F

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.444 (lit.)

bp

179 °C/15 mmHg (lit.)
314.4 °C±5.0 °C/760 mmHg (lit.)

mp

14-16 °C (lit.)

density

0.789 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCCC=C

InChI

1S/C18H36/c1-3-5-7-9-11-13-15-17-18-16-14-12-10-8-6-4-2/h3H,1,4-18H2,2H3

InChI key

CCCMONHAUSKTEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

300.2 °F - closed cup

Flash Point(C)

149.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaping Du et al.
Journal of the American Chemical Society, 132(5), 1470-1471 (2010-01-19)
Monodisperse Ag(2)S quantum dots (QDs) were synthesized via pyrolysis of Ag(DDTC) in oleic acid, octadecylamine, and 1-octadecene. The uniform alkyl-capped Ag(2)S QDs with a size of 10.2 nm emit near-IR emission at 1058 nm under 785 nm excitation.
Wenwen Zhou et al.
Nanotechnology, 22(19), 195607-195607 (2011-03-26)
CuInSe(2) (CIS) nanodandelion structures were synthesized by a two-step solvothermal approach. First, InSe nanodandelions were prepared by reacting In(acac)(3) with trioctylphosphine-selenide (TOP-Se) in 1-octadecene (ODE) at 170 °C in the presence of oleic acid. These InSe dandelions were composed of polycrystalline
Zheng Li et al.
Journal of the American Chemical Society, 133(43), 17248-17256 (2011-09-24)
Formation of CdS nanocrystals in the classic approach (with octadecene (ODE) as the solvent and elemental sulfur and cadmium carboxylate as the precursors) was found to be kinetically dependent on reduction of elemental sulfur by ODE, which possessed a critical
Synthesis, shape control, and optical properties of hybrid Au/Fe3O(4) "nanoflowers".
Yanhu Wei et al.
Small (Weinheim an der Bergstrasse, Germany), 4(10), 1635-1639 (2008-07-19)
Eun-Jung Park et al.
Archives of toxicology, 88(8), 1607-1618 (2014-02-15)
The unique properties of nanoparticles and biological systems are important factors affecting the biological response following nanoparticle exposure. Iron oxide nanoparticles are classified mainly as magnetite (M-FeNPs) and maghemite (NM-FeNPs). In our previous study, NM-FeNPs induced autophagic cell death in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service