Skip to Content
Merck
All Photos(1)

Key Documents

43306

Supelco

Phytanic acid

mixture of isomers, analytical standard

Synonym(s):

3,7,11,15-Tetramethylhexadecanoic acid, NSC 108698, Phytanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€169.00
100 MG
€510.00

€169.00


Please contact Customer Service for Availability


Select a Size

Change View
10 MG
€169.00
100 MG
€510.00

About This Item

Empirical Formula (Hill Notation):
C20H40O2
CAS Number:
Molecular Weight:
312.53
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.24

€169.00


Please contact Customer Service for Availability

grade

analytical standard

quality

mixture of isomers

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

SMILES string

CC(C)CCCC(C)CCCC(C)CCCC(C)CC(O)=O

InChI

1S/C20H40O2/c1-16(2)9-6-10-17(3)11-7-12-18(4)13-8-14-19(5)15-20(21)22/h16-19H,6-15H2,1-5H3,(H,21,22)

InChI key

RLCKHJSFHOZMDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service