Skip to Content
Merck
All Photos(1)

Key Documents

36372

Supelco

Tebufenpyrad

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

4-Chloro-N-(4-tert-butylbenzyl)-3-ethyl-1-methyl-1H-pyrazole-5-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€71.00

€71.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€71.00

About This Item

Empirical Formula (Hill Notation):
C18H24ClN3O
CAS Number:
Molecular Weight:
333.86
Beilstein:
8636471
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€71.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

CCc1nn(C)c(C(=O)NCc2ccc(cc2)C(C)(C)C)c1Cl

InChI

1S/C18H24ClN3O/c1-6-14-15(19)16(22(5)21-14)17(23)20-11-12-7-9-13(10-8-12)18(2,3)4/h7-10H,6,11H2,1-5H3,(H,20,23)

InChI key

ZZYSLNWGKKDOML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Tebufenpyrad is a non-systemic pyrazolecarboxamide acaricide that gets activated both by contact and ingestion. It targets the NADH-ubiquinone oxidoreductase (complex I) site and acts as a mitochondrial electron transport inhibitor (MET I). Tebufenpyrad is used for the protection of all stages of Tetranychus, Panonychus, Oligonychus, and Eotetranychus spp. in top fruit, vines, citrus, vegetables, hops, ornamentals, melons, and cotton.
According to the Commission Implementing Regulation (EU) No 2009/11/EC of 18th February 2009, tebufenpyrad is not approved for use in the European Union (EU) as a plant protection agent, following the Regulation (EC) No. 1107/2009. But maximum residue levels of 0.01 mg/kg to 1.5 mg/kg are allowed for the presence of tebufenpyrad in products of plant and animal origin, according to the EU Reg. No 2017/693.
Tebufenpyrad has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Profenofos CRM may also be used as given below:
  • Evaluation of 250 pesticide residues extracted by QuEChERS method from 103 processed fruit samples using ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
  • Gas chromatography-tandem mass spectrometry (GC-MS/MS) based analysis of 213 pesticide residues in soybean, after their extraction by QuEChERS procedure
  • Detection of 252 pesticides extracted from surface water samples by solid-phase extraction (SPE) using liquid chromatography quadrupole-orbitrap high-resolution tandem mass spectrometry
  • Multi-residue analysis of 160 multiclass pesticides using ethyl acetate (EtAC)/n. hexane mixture for extraction from chamomile, thyme, and marjoram samples and their analysis by gas chromatography-tandem mass spectrometry (GC-MS/MS)
  • Simultaneous determination of 34 pesticides in various environmental water samples by dispersive liquid-liquid microextraction (DLLME) technique in combination with gas chromatography-mass spectrometry(GC-MS/MS)
  • Quantitation of 40 multi-class insecticides and fungicides in 81 apple samples by using gas- and liquid- chromatography-mass, following a QuEChERS-based extraction

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service