Skip to Content
Merck
All Photos(1)

Key Documents

36003

Supelco

Oxydemeton-methyl

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Demeton-S-methyl sulfoxide, S-[2-(Ethylsulfinyl)ethyl] O,O-dimethyl phosphorothioate, Methyl oxydemeton

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€257.00

€257.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€257.00

About This Item

Empirical Formula (Hill Notation):
C6H15O4PS2
CAS Number:
Molecular Weight:
246.28
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€257.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

−20°C

SMILES string

[P](=O)(SCC[S+]([O-])CC)(OC)OC

InChI

1S/C6H15O4PS2/c1-4-13(8)6-5-12-11(7,9-2)10-3/h4-6H2,1-3H3

InChI key

PMCVMORKVPSKHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service