Skip to Content
Merck
All Photos(1)

Key Documents

33039

Supelco

HMDS+TMCS+Pyridine, 3:1:9 (Sylon HTP)

pkg of 25 mL

Synonym(s):

Chlorotrimethylsilane, Hexamethyldisilazane, Trimethylchlorosilane, Trimethylsilyl chloride, 1,1,1,3,3,3–Hexamethyldisilane+Τrimethylchlorosilane+Pyridine, Sylon HTP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

grade

for GC derivatization

description

3:1:9 (SYLON HTP)

composition

1,1,3,3,3-Hexamethyldisilazane, 22.9%
Pyridine, 69.4%
Trimethylchlorosilane, 7.7 % (w/v)

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

packaging

pkg of 25 mL

Looking for similar products? Visit Product Comparison Guide

Application

HMDS+TMCS+Pyridine in the ratio of 3:1:9 may be used as the silylating agent, during multi-targeted profiling of the major phytohormones by gas chromatography-mass spectrometry (GC-MS). This reagent was used as a derivatizing agent during sample processing during determination of 5-hydroxymethylfurfural in wood using GC-MS method.

Legal Information

Sylon is a trademark of Sigma-Aldrich Co. LLC
Sylon CT is a trademark of Sigma-Aldrich Co. LLC

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a GC-MS method for 5-hydroxymethylfurfural determination in wood after steam-explosion pretreatment.
Senila, Lacrimioara, et al.
Revista de Chimie (Bucharest, Romania), 63, 559-563 (2012)
Mass spectra of trimethylsilyl derivatives of homogeneous cerebrosides (monoglycosylceramides).
K A Karlsson et al.
Chemistry and physics of lipids, 9(3), 230-246 (1972-11-01)
The measurement of myo-inositol, myo-inosose-2 and scyllo-inositol in mammalian tissues.
W R Sherman et al.
Biochimica et biophysica acta, 158(2), 197-205 (1968-05-01)
T.Nambara et al.
Journal of Chromatography A, 34, 526-526 (1968)
Studies on sphingosines. 10. Use of trimethylsilyl ethers for the gas chromatography and mass spectrometry sphingosines.
K A Karlsson
Acta chemica Scandinavica, 19(10), 2425-2427 (1965-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service