27505-U
Vials, screw top, graduated, solid cap, preassembled
volume 2 mL, clear glass vial, thread for 8-425, PTFE liner
About This Item
Recommended Products
material
PTFE liner
black phenolic solid cap
clear glass vial
feature
closure type black phenolic solid top cap
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H × I.D.
12 mm × 32 mm × 4.6 mm
volume
2 mL
fitting
thread for 8-425
Looking for similar products? Visit Product Comparison Guide
General description
related product
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service