27101
Septa, PTFE/red rubber, pkg 100
PTFE/red rubber septum, seal diam. 11 mm, pkg of 100 ea, for use with 2mL-2.5mL standard opening crimp top vials
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
PTFE septum face
PTFE/red rubber
PTFE/red rubber septum
packaging
pkg of 100 ea
O.D.
11 mm
seal diam.
11 mm
compatibility
for use with 2mL-2.5mL standard opening crimp top vials
Looking for similar products? Visit Product Comparison Guide
General description
Septa are selected based on the chemical properties of the sample and the temperature to which the vial will be subjected during the analysis. PTFE/Rubber septa are used with most organic solvents. These must not be used with aromatics, carbon disulfides, chlorinated solvents, and hydrocarbons.
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service