Skip to Content
Merck
All Photos(1)

Documents

27052

Supelco

Screw cap (open-top), polypropylene

black polypropylene cap, thread for 8-425, for use with 2 mL vial (standard opening), pkg of 100 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121821

material

black polypropylene cap

feature

autoclavable
closure type black polypropylene hole cap

packaging

pkg of 100 ea

parameter

135 °C temp. limit

fitting

thread for 8-425

compatibility

for use with 2 mL vial (standard opening)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

These hole caps do not come with septa.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adsorption of cationic peptides to solid surfaces of glass and plastic.
Kristensen, Kasper, Jonas R. Henriksen, and Thomas L. Andresen.
PLoS ONE, 10.5, e0122419-e0122419 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service