Skip to Content
Merck
All Photos(1)

Documents

10341

Supelco

Amberlite IRN150 hydrogen and hydroxide form

hydrogen and hydroxide form, matrix styrene/divinylbenzene (gel), 22-26 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817

grade

nuclear grade

description

cross-linkage 8%

form

beads

packaging

bucket of 500 g

parameter

60 °C max. temp.

moisture

49-55% (cation)
54-60% (anion)

technique(s)

LPLC: suitable

matrix

styrene/divinylbenzene (gel)

matrix active group

mixed mode

particle size

22-26 mesh
580-680 μm (anion exchanger)
600-700 μm (cation exchanger)

operating pH

0-14

capacity

1.2 meq/g by dry weight(anion)
1.9 meq/g by dry weight(cation)

separation technique

mixed mode

Looking for similar products? Visit Product Comparison Guide

General description

Amberlite IRN150 is a strong base anion and cation exchange mixed bed resin.
Mixed bed resin for primary water chemistry control in once-through systems. Useful in industrial water treatment where as-supplied resin must have absolute minimum of ionic and non-ionic contamination.

Application

Amberlite IRN150 was used to determine the particle size distribution using photo correlation spectroscopy.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study on the Correlation between Lateral Diffusion Effect and Effective Charge in Neutral Liposomes
Galera-Cortes, Elisa, et al.
Langmuir, 26, 2665-2670 (2009)
Comparison of strongly basic anion exchange resins applicability for the removal of palladium (II) ions from acidic solutions
Wolowicz, A., and Z. Hubicki
Chemical Engineering Journal, 171, 206-215 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service