Skip to Content
Merck
All Photos(8)

Key Documents

ZRB2144

Sigma-Aldrich

Anti-ZAP-70 Antibody, clone 1D9 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

70 kDa zeta-chain associated protein, EC:2.7.10.2, Syk-related tyrosine kinase, Tyrosine-protein kinase ZAP-70

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D9, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1D9 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Tyrosine-protein kinase ZAP-70 (ZAP-70). It targets an epitope within 17 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the internal region of human Tyrosine-protein kinase ZAP-70 (ZAP-70).

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell plasma membrane preparation.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ZAP-70 in Jurkat cell plasma membrane preparation.

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound ZAP-70 peptide with a KD of 3.9 x 10-7 in an affinity binding assay.

Flow Cytometry Analysis: 0.1 µg from a representative lot detected ZAP-70 in one million Jurkat cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected ZAP-70 in Human tonsil tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Tyrosine-protein kinase ZAP-70 (UniProt: P43403; also known as EC:2.7.10.2, 70 kDa zeta-chain associated protein, Syk-related tyrosine kinase) is encoded by the ZAP70 (also known as SRK) gene (Gene ID: 7535) in human. ZAP-70 is a tyrosine kinase that is expressed in T cells and NK cells and plays an essential role in regulation of the adaptive immune response. Its expression is also observed in early thymocytes and pro/pre-B-cells. It is involved in regulating motility, adhesion, and cytokine expression of mature T-cells and thymocyte development.  In quiescent T cells, it is cytoplasmic and upon TCR activation, it is recruited at the plasma membrane by interacting with CD247/CD3Z.  ZAP-70 contains two tandem SH2 domains (aa 10-102 and 163-254) that bind to the doubly phosphorylated tyrosine-based activation motif (ITAM) of CD247/CD3Z and the non-canonical phosphorylated tyrosine-based activation motif (TAM) of Ras Homolog Family Member H (RHOH). Its protein kinase domain is localized in amino acids 338-600. It has an interdomain B region (aa 255-337), located between the second SH2 and the kinase domain, contains 3 tyrosine residues (Tyr-292, 315, 319) that are phosphorylated following TCR activation. Phosphorylation of Tyr-315 and Tyr-319 is reported to be essential for ZAP70 positive function on T-lymphocyte activation, whereas Tyr-292 is shown to have a negative regulatory role. Within the protein kinase domain, Tyr-492 and Tyr-493 are also shown to be phosphorylated after TCR induction. Here Tyr-492 plays a negative regulatory role and Tyr-493 has a positive role and can be  dephosphorylated by protein tyrosine phosphatase non-receptor type 22 (PTN22). Aberrant high expression of ZAP-70 has been reported in a subset of Chronic Lymphocytic Leukemia (CLL) subjects and reflects an unfavorable clinical outcome. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Chen, J., et al.(2020). Front. Oncol. 10; 595832; Fischer, A., et al. (2010). Semin. Immunopathol. 32(2); 107-116; Chan, AC., et al. (1992). Cell. 71(4); 649-662).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service