Skip to Content
Merck
All Photos(10)

Key Documents

ZRB1949

Sigma-Aldrich

Anti-STAT6 Antibody, clone 1C13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Signal transducer and activator of transcription 6;IL-4 Stat

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€194.00
4 X 25 μL
€399.00

€194.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€194.00
4 X 25 μL
€399.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€194.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C13, recombinant monoclonal

description

1C13 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 94.14 kDa
observed mol wt ~105 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... STAT6(6778)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C13 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects STAT6. It targets an epitope within 25 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the N-terminal region of human STAT6.

Application

Quality Control Testing

Evaluated by Western Blotting in HepG2 cell lysates.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected STAT6 in HepG2 cell lysates.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected STAT6 in human fallopian tube tissue sections.

Flow Cytometry Analysis: 1 μg from a representative lot detected STAT6 in one million HepG2 cells.

Affinity Binding Assay: A representative lot of this antibody bound STAT6 peptide with a KD of 1.9 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Signal transducer and activator of transcription 6 (UniProt: P42226; also known as IL-4 Stat) is encoded by the STAT6 gene (Gene ID: 6778) in human. The Janus kinase (JAK)/signal transducers and activators of transcription (STAT) signaling pathway play an important role in cell proliferation, cell differentiation, cell migration, and cell death. It is the principal signaling mechanism for a variety of cytokines and growth factors. Constitutive activation or dysregulation of JAK/STAT signaling can result in inflammatory disease, erythrocytosis, gigantism, and leukemia. In mammalian cells seven different STATs been recognized. They contain a conserved tyrosine residue near the C-terminus, which is phosphorylated by JAKs. This tyrosine phosphorylation allows the dimerization of STATs by interacting with a conserved SH2 domain. In their latent form STATs are located in the cytoplasm and upon phosphorylation they are transported into the nucleus by an importin-dependent mechanism. In the nucleus, dimerized STATs bind to specific regulatory sequences to activate or repress transcription of target genes. Activated STATs are reported to stimulate transcription of the SOCS genes and the resulting SOCS proteins bind to phosphorylated JAKs and block their activity. The SOCS can also bind to phosphotyrosines on the receptors and block the recruitment of STATs to these receptors. STAT6 is considered to be the key to the IL-4 signaling pathway and a driver of Th2 cell differentiation. It carries out a dual function: signal transduction and activation of transcription. It is involved in several distinct aspects of allergic inflammatory disease, such as airway hyperresponsiveness, eosinophilic infiltration, and responses of mast cells. STAT6-deficient mice are reported to lack most of the physiologic functions associated with IL-4, in particular the ability of IL-4 to induce the in vitro differentiation of Th2 cells. Three isoforms of STAT6 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Chu-K-H., et al. (2021). Front. Immunol. 11; 615868; Lu, X., et al. (2007). Mol. Cell Biol. 27(6); 2166-2179).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service