Z744322
Wilmad® NMR Reaction Monitoring System
tube O.D. 5 mm, tube L 7 in., pack of 1 ea
Synonym(s):
NMR monitoring tube, NMR reaction tube
About This Item
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Wilmad WG-RMS-7
tube L
7 in.
tube O.D.
5 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- In-tube mixing
- Before and after reaction picture
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service