Z741615
Prospenser Bottle-top Dispenser
dispensing volume 0.2-10 mL
All Photos(1)
About This Item
UNSPSC Code:
41121500
Recommended Products
manufacturer/tradename
Sartorius 723051
dispensing volume measuring range
0.2-10 mL
General description
The Prospenser bottle-top dispensers deliver safe and precise dispensing of liquids, including strong acids, bases and solvents from a supply bottle. Special features include an easy-to-adjust calibration mechanism and anti-drip precision valve for enhanced accuracy and usability. A fine adjustment mechanism allows for even greater reproducibility for repeat dispensing. Prospensers are fully autoclavable and easy to dismantle for cleaning and maintenance. Wide range of adapters are included to fit most common bottle sizes. All models are fitted with an anti-drip safety valve or a closure cap, a feature which guards against drips, when the bottle-top dispensers are not in use. A fine adjustment mechanism allows for even greater reproducibility for repeat dispensing. Wide range of adapters included to fit most common bottle sizes. Fully autoclavable at 121o C, 2 bar for 20 minutes.The Prospenser bottle-top dispensers deliver safe and precise dispensing of liquids, including strong acids, bases and solvents from a supply bottle. Special features include an easy-to-adjust calibration mechanism and anti-drip precision valve for enhanced accuracy and usability. A fine adjustment mechanism allows for even greater reproducibility for repeat dispensing. Prospensers are fully autoclavable and easy to dismantle for cleaning and maintenance. Wide range of adapters are included to fit most common bottle sizes. All models are fitted with an anti-drip safety valve or a closure cap, a feature which guards against drips, when the bottle-top dispensers are not in use. A fine adjustment mechanism allows for even greater reproducibility for repeat dispensing. Wide range of adapters included to fit most common bottle sizes. Fully autoclavable at 121o C, 2 bar for 20 minutes.
Choose from one of the most recent versions:
Certificates of Analysis (COA)