Z740460
Isohelix Spin+Collect™ devices
pkg of 150 ea (5 per bag)
Select a Size
€68.60
Select a Size
About This Item
€68.60
Recommended Products
sterility
non-sterile
packaging
pkg of 150 ea (5 per bag)
manufacturer/tradename
Isohelix SC/ST-150
tube size
1.5 mL
working volume
1.5 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features:
- Yields increase over 25%
- Recovers sample deeply embedded in the matrix
- Requires a single centrifuge step
- Disposable, sterile and packaged aseptically
- Tested for Human DNA contamination
Additional info:
Qty 100 x Spin+Collect devices packed 5 per bag with 100 x 1.5 mL matched tubes.
Qty 150 x Spin+Collect devices packed 5 per bag.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service