Skip to Content
Merck
All Photos(1)

Documents

Z740251

AluminaSeal for cold and PCR storage

sterile

Synonym(s):

Al plate seal, aluminium plate seal, storage plate seal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.22

material

aluminum

sterility

sterile

feature

piercable

packaging

pkg of 100 ea (sheets)

manufacturer/tradename

Diversified Biotech ALUM-1000

technique(s)

PCR: suitable

General description

  • Ideal for cold storage, moderate and high temperature incubation, protection of light sensitive samples and high throughput samples
  • Strong 2.7 mil thick pierceable aluminum film
  • Non-cytotoxic and sterile
  • Temperature stable from -80°C to 110°C
  • Resistant to DMSO, chemicals, solvents and other common reagents
  • Two scored end tabs for easy positioning

Legal Information

AluminaSeal is a trademark of Diversified Biotech

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service