Z721727
KDS Continuous cycle syringe pump, series 270
AC/DC input 110 V AC
Synonym(s):
dosing pump, infusion pump, pump infusion
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
2 channels
manufacturer/tradename
KD Scientific 780270-110V
availability
not available in EU
technique(s)
HPLC: suitable
cell culture | mammalian: suitable
volume
0.01-60 mL
suitability
suitable for infusion
Looking for similar products? Visit Product Comparison Guide
General description
Continuous Cycle Syringe Pump. The KDS270 can hold up to four syringes and can cycle continuously back and forth in a push-pull action. As two syringes are infusing, two syringes are withdrawing at the same rate. At the end of the set volume the direction is automatically reversed and the next cycle begins. With the use of 3-way valves the pump can empty and refill syringes in a continuous dispense.
- Holds four syringes, 10 μL to 60 mL each. With larger syringes the full volume may not be useable. [60 mL syr - 40 mL useable, 30 mL syr - full]
- Backlit LCD display
- Knob locks/unlocks drive block for effortless, drag-free adjustment
- Simple menu-driven setup
- Select syringe size from displayed table
- Dispense volume
- Dispense flow rate
- Continuous dispense volume display
- Preset volume control and automatic shut-off
- Settings can be reviewed or changed during operation
- Stall detection
- Choice of unit selection
- Last settings stored in permanent memory
- Built-in RS232C interface for computer "daisy chaining" up to 100 pumps
- TTL interface for foot switch, timer, relay control, outputs for run indicator, valve control
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service