Documents
Z637475
BRAND® microtube cap inserts, PP
red
About This Item
UNSPSC Code:
41121702
NACRES:
NB.11
manufacturer/tradename
BRAND 780722
color
red
General description
The BRAND® Microtube Cap Inserts are designed for use with microtubes featuring attached or bulk screw caps that are non-tamper evident. Made from durable polypropylene (PP), these inserts provide a secure closure for sample integrity. They are suitable for a wide range of applications, with an operating temperature range from -196 °C to +121 °C, making them ideal for both cryogenic and high-temperature environments. These cap inserts are essential for ensuring reliable storage and handling of samples in various laboratory settings.
Other Notes
BR780722 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG