Z382077
Nalgene® Lowboy carboy with spigot
nominal capacity 8 L (2 gal), polypropylene copolymer bottle (PPCO)
Synonym(s):
carboys
All Photos(1)
About This Item
Recommended Products
material
polypropylene copolymer bottle (PPCO)
polypropylene spigot and screw closure
description
Not recommended for use with most organic solvents
manufacturer/tradename
Nalgene® 2324 style
L × W × H
6 1/2 in. (165 mm) × 14 1/2 in. (368 mm) × 12 in. (305 mm)
nominal capacity
8 L (2 gal)
Related Categories
General description
Fits into tight places and can be stacked three high. Integral handgrip for safe and easy transport. Tolerant of strong acids and bases.
Similar to Style 2323, but all polypropylene.
Chem/Phys Resistance
Autoclavable
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries