Z374563
Eppendorf® Multipette® Plus pipette
Synonym(s):
Adjustable volume pipette
All Photos(1)
About This Item
Recommended Products
feature
electronic: no
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022260201
Eppendorf® 4981 000.019
General description
Accepts any of seven interchangeable combitips; each tip gives a choice of 20 preset volumes. The combitip′s integral reservoir lets you pipette up to 100 times without refilling. Pipette automatically recognizes tip and digitally displays selected volume.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Multipette is a registered trademark of Eppendorf AG