Z330086
BRAND® wide-mouth bottles, LDPE
capacity 50 mL, with cap
Synonym(s):
ldpe bottle, polyethylene bottles, polyethylene containers
About This Item
Recommended Products
material
low-density polyethylene bottle
round bottle
translucent bottle
wide-mouth bottle
Joint
Joints threaded neck (GL32)
feature
with cap
packaging
pkg of 100 ea
manufacturer/tradename
BRAND 129628
capacity
50 mL
diam. × H
38 mm × 80 mm
color
natural
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service