biological source
Saccharomyces cerevisiae
Quality Level
form
powder
solubility
H2O: 1%, hazy
application(s)
microbiology
InChI
1S/C19H14O2/c20-18(21)11-17-15-8-4-3-7-14(15)16-10-9-12-5-1-2-6-13(12)19(16)17/h1-10,17H,11H2,(H,20,21)
InChI key
GQNBDGXKDJSVGQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Yeast extract is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications.
Other Notes
Water soluble portion of autolyzed yeast with intact B-complex vitamins. Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates and can be used as additive for culture media.
Caution
may contain black specks
Preparation Note
Spray dried, autolyzed yeast extract.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service