V8004
L-Valine-UL-14C hydrochloride
≥98% (Radiochemical Purity, HPLC), aqueous ethanol solution
Synonym(s):
(S)-(+)-Valine-UL-14C
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
[14CH3]214CH14CH[NH2]14COOH
CAS Number:
Molecular Weight:
117.15
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
≥98% (Radiochemical Purity, HPLC)
form
aqueous ethanol solution
extent of labeling
200-300 mCi per mmol
storage temp.
2-8°C
SMILES string
[14CH3][14CH]([14CH3])C(N)[14C](O)=O
Looking for similar products? Visit Product Comparison Guide
Packaging
Combi-vial
Physical form
Aqueous solution containing 2% ethanol
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service