Skip to Content
Merck
All Photos(1)

Key Documents

UC322

Sigma-Aldrich

Naloxone-3-glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H29NO10
CAS Number:
Molecular Weight:
503.50
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

color

white

mp

200-205 °C

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C(O)=O)Oc2ccc3C[C@H]4N(CC[C@@]56[C@@H](Oc2c35)C(=O)CC[C@@]46O)CC=C

InChI

1S/C25H29NO10/c1-2-8-26-9-7-24-15-11-3-4-13(34-23-18(30)16(28)17(29)20(36-23)22(31)32)19(15)35-21(24)12(27)5-6-25(24,33)14(26)10-11/h2-4,14,16-18,20-21,23,28-30,33H,1,5-10H2,(H,31,32)/t14-,16+,17+,18-,20+,21+,23-,24+,25-/m1/s1

InChI key

DSRNCSJRHACUJL-JXOQMJNJSA-N

Application

Naloxone-3-glucuronide can be used for the assay of naloxone metabolites.

Biochem/physiol Actions

Phase II metabolite of naloxone.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service